Learn More
Sampling Systems™ Silicone Seal for Storage Containers
These seals when used with toggle clamps will make storage containers leak-proof. Choose the right sized seal for your container.
486.00 NOK - 692.00 NOK
Specifications
Material | Silicone |
---|---|
Type | Seal, O-ring |
For Use With (Application) | Leak-proof sealing |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15829731
|
Sampling Systems™
SIL-20-FDA |
20L storage container |
492.00 NOK
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
15839731
|
Sampling Systems™
SIL-25-FDA |
25L storage container |
492.00 NOK
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
15859731
|
Sampling Systems™
SIL-30-FDA |
30L storage container |
486.00 NOK
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
15869731
|
Sampling Systems™
SIL-50-FDA |
50L storage container |
486.00 NOK
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
15889731
|
Sampling Systems™
SIL-75-FDA |
75L storage container |
687.00 NOK
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
15889721
|
Sampling Systems™
SIL-100-FDA |
100L storage container |
692.00 NOK
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
Description
- O-ring design
- Made from FDA-acceptable grade silicone
- Suitable for use with both 304 and 316L stainless steel storage containers
- Wide range of sizes available
Specifications
Silicone | |
Seal, O-ring | |
Leak-proof sealing |