Learn More
Sampling Systems™ Silicone Seal for Storage Containers
These seals when used with toggle clamps will make storage containers leak-proof. Choose the right sized seal for your container.
Brand: Sampling Systems™ SIL-30-FDA
This item is not returnable.
View return policy
Description
- O-ring design
- Made from FDA-acceptable grade silicone
- Suitable for use with both 304 and 316L stainless steel storage containers
- Wide range of sizes available
Specifications
Leak-proof sealing | |
30L storage container | |
Silicone | |
Seal, O-ring |