Learn More
Buerkle™ Closing Cap with Inner Thread
Manufactured from polypropylene.
51.63 NOK - 64.88 NOK
Specifications
Autoclavable | Autoclavable |
---|---|
Material | Polypropylene |
Product Type | Closing Cap |
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thread Size | Price | Quantity & Availability | |||||
10044990
|
Buerkle™
8535-0012 |
1/2 in. |
51.63 NOK
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
|||||
10370391
|
Buerkle™
8535-0034 |
3/4 in. |
64.88 NOK
Each |
Estimated Shipment: 17-09-2025
Log in to see stock. |
|||||
Description
- Polypropylene construction
- Autoclavable to 121°C
- Safe for use with foodstuffs
- Supplied in packs of 10
Specifications
Autoclavable | |
Polypropylene | |
Closing Cap |