Learn More
Buerkle™ Closing Cap with Inner Thread
Manufactured from polypropylene.
Brand:  Buerkle™ 8535-0012
46.41 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10044990
  • 51.63 NOK / Each
Only null left
Estimated Shipment: 04-09-2025
Description
  • Polypropylene construction
  • Autoclavable to 121°C
  • Safe for use with foodstuffs
  • Supplied in packs of 10
Specifications
Specifications
EU Foodstuffs
Polypropylene
Closing Cap
Autoclavable
1/2 in.