Learn More
Buerkle™ Closing Cap with Inner Thread
Manufactured from polypropylene.
Brand: Buerkle™ 8535-0034
58.40 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Polypropylene construction
- Autoclavable to 121°C
- Safe for use with foodstuffs
- Supplied in packs of 10
Specifications
Autoclavable | |
Polypropylene | |
3/4 in. | |
Closing Cap |