Learn More
Buerkle™ Nipple with Outer Thread
Manufactured from polypropylene.
41.55 NOK - 47.68 NOK
Specifications
Certifications/Compliance | EU Foodstuffs |
---|---|
Autoclavable | Autoclavable |
Material | Polypropylene |
Product Type | Nipple Outer Thread |
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
10558082
|
Buerkle™
8505-0012 |
13.5 mm |
41.55 NOK
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
|||||
10709861
|
Buerkle™
8505-0034 |
18 mm |
47.68 NOK
Each |
Estimated Shipment: 24-07-2025
Log in to see stock. |
|||||
Description
- Polypropylene construction
- Autoclavable to 121°C
- Safe for use with foodstuffs
- Supplied in packs of 10
Specifications
EU Foodstuffs | |
Autoclavable | |
Polypropylene | |
Nipple Outer Thread |