Learn More
Buerkle™ Nipple with Outer Thread
Manufactured from polypropylene.
Brand: Buerkle™ 8505-0034
This item is not returnable.
View return policy
Description
- Polypropylene construction
- Autoclavable to 121°C
- Safe for use with foodstuffs
- Supplied in packs of 10
Specifications
EU Foodstuffs | |
Polypropylene | |
18 mm |
Autoclavable | |
3/4 in. | |
Nipple Outer Thread |