Learn More
Buerkle™ Nipple with Outer Thread
Manufactured from polypropylene.
Brand: Buerkle™ 8505-0012
This item is not returnable.
View return policy
Description
- Polypropylene construction
- Autoclavable to 121°C
- Safe for use with foodstuffs
- Supplied in packs of 10
Specifications
EU Foodstuffs | |
Polypropylene | |
13.5 mm |
Autoclavable | |
1/2 in. | |
Nipple Outer Thread |
Product Content Correction
Product Title
Buerkle™ Nipple with Outer Thread > Width: 13.5mm