Learn More
SHOWA 707HVO Orange Nitrile Gloves
The 707HVO gloves are highly tactile and fit like a second skin, all whilst protecting against grease, chemicals, and abrasion.
238.00 NOK
Specifications
Certifications/Compliance (Europe) | EN 388:2016, EN ISO 374-1:2016/Type A, EN ISO 374-5:2016 |
---|---|
Color | Orange |
Glove Cuff Style | Rolled |
Model | 707HVO |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17897311
|
SHOWA
707HVO-06XS |
6 | N/A | ||||||
17807321
|
SHOWA
707HVO-07S |
7 | N/A | ||||||
17817321
|
SHOWA
707HVO-08M |
8 | N/A | ||||||
17827321
|
SHOWA
707HVO-09L |
9 | N/A | ||||||
17837321
|
SHOWA
707HVO-10XL |
10 | N/A | ||||||
17847321
|
SHOWA
707HVO-11XXL |
11 | N/A | ||||||
Description
- Fluorescent
- Lightweight
- Chlorinated
- Eco Best Technology (EBT)
- Unlined
- Unsupported
Specifications
EN 388:2016, EN ISO 374-1:2016/Type A, EN ISO 374-5:2016 | |
Orange | |
Rolled | |
707HVO |