Learn More
SHOWA 707HVO Orange Nitrile Gloves
Click to view available options
Size:
6
7
8
9
10
11
Description
- Fluorescent
- Lightweight
- Chlorinated
- Eco Best Technology (EBT)
- Unlined
- Unsupported
Specifications
Specifications
Certifications/Compliance (Europe) | EN 388:2016, EN ISO 374-1:2016/Type A, EN ISO 374-5:2016 |
Color | Orange |
Size | 10 |
Glove Cuff Style | Rolled |
Model | 707HVO |