Learn More
SHOWA 707HVO Orange Nitrile Gloves
Product Code. 17817321
Click to view available options
Size:
6
7
8
9
10
11
The 707HVO gloves are highly tactile and fit like a second skin, all whilst protecting against grease, chemicals, and abrasion.
  • Fluorescent
  • Lightweight
  • Chlorinated
  • Eco Best Technology (EBT)
  • Unlined
  • Unsupported
Specifications
Certifications/Compliance (Europe) EN 388:2016, EN ISO 374-1:2016/Type A, EN ISO 374-5:2016
Color Orange
Size 8
Glove Cuff Style Rolled
Model 707HVO