Learn More
Macherey-Nagel™ Buffer NT3 Concentrate
Concentrated ethanolic wash buffer for removing contaminations from bound DNA when using NucleoSpin™ and NucleoTraP™ PCR Clean-up kits. To prepare working buffer solution, add 25 mL concentrate to 100 mL ethanol (yields 125 mL working solution).
Brand: Macherey-Nagel™ 740598.1
157.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
NucleoTraP™ and NucleoSpin™ PCR clean-up kits allow DNA fragments to be purified from enzymatic reactions, such as PCR, as well as from agarose gels. Compatible kits include:
- NucleoSpin™ Gel and PCR Clean-up (standard, Midi, Maxi)
- NucleoSpin™ 8/96 PCR Clean-up (full and core kit)
- NucleoTraP™ CR and NucleoTraP™
Specifications
Colorless | |
NucleoTraP™ and NucleoSpin™ PCR clean-up kits | |
200 mL | |
Odorless | |
Liquid | |
Wash buffer |
5X (50 mL concentrate yields 125 mL working solution) | |
7.0 to 8.0 | |
Concentrated | |
Room Temperature (18°C to 25°C) | |
Purification (clean up) of DNA |