Learn More
VAV2 Mouse anti-Human, Unconjugated, Clone: 3F11B5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67108-1-IG-150UL
 View more versions of this product
Product Code. 16859545
  • 4652.90 SEK / 150µL
Description
The VAV2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human VAV2. This antibody recognizes human, pig, mouse, rat antigen. The VAV2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
VAV2
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, ELISA
3F11B5
Unconjugated
150 μL
7410
IgG2a