missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UPF1 Rabbit, anti-Human, Clone: 3O15, Proteintech
Rabbit Recombinant Monoclonal Antibody
1743.12 NOK - 5135.76 NOK
Specifications
Antigen | UPF1 |
---|---|
Clone | 3O15 |
Concentration | 300 μg/mL |
Applications | Western Blot, Immunofluorescence, ELISA |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18047085
|
Proteintech
82791-1-RR-100UL |
100 μL |
5135.76 NOK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18089455
|
Proteintech
82791-1-RR-20UL |
20 μL |
1743.12 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The UPF1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human UPF1. This antibody recognizes human, rat antigen. The UPF1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
UPF1 | |
300 μg/mL | |
Liquid | |
Human, Rat | |
ATP dependent helicase RENT1, FLJ43809, FLJ46894, HUPF1, KIAA0221, NORF1, pNORF1, RENT1, UPF1 | |
Primary |
3O15 | |
Western Blot, Immunofluorescence, ELISA | |
Rabbit | |
5976 | |
IgG | |
Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title