Learn More
UPF1 Rabbit, anti-Human, Clone: 3O15, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82791-1-RR-100UL
Description
The UPF1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human UPF1. This antibody recognizes human, rat antigen. The UPF1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
UPF1 | |
3O15 | |
ATP dependent helicase RENT1, FLJ43809, FLJ46894, HUPF1, KIAA0221, NORF1, pNORF1, RENT1, UPF1 | |
100 μL | |
5976 | |
Store at -20°C. Stable for one year after shipment. | |
Liquid |
Western Blot, Immunofluorescence, ELISA | |
300 μg/mL | |
Rabbit | |
Primary | |
Human, Rat | |
150 kDa | |
IgG |