Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML E1
Adjusted to Meet or Exceed OIML E1 Tolerances
1880.00 NOK - 50550.00 NOK
Specifications
Certifications/Compliance | No Certificate |
---|---|
Material | Troemner Alloy 8 |
Includes | Protective Case |
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Mass | Price | Quantity & Availability | |||||
16642312
|
Ohaus™
30517972 |
1 g |
1880.00 NOK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||||
16652312
|
Ohaus™
30517973 |
2 g |
1925.00 NOK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||||
16662312
|
Ohaus™
30517974 |
5 g |
1965.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
16672312
|
Ohaus™
30517975 |
10 g |
2150.00 NOK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||||
16682312
|
Ohaus™
30517976 |
20 g |
2255.00 NOK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||||
16692312
|
Ohaus™
30517977 |
50 g |
2695.00 NOK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||||
16602322
|
Ohaus™
30517978 |
100 g |
3195.00 NOK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||||
16612322
|
Ohaus™
30517979 |
200 mg |
3770.00 NOK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||||
16622322
|
Ohaus™
30517980 |
500 g |
5325.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
16632322
|
Ohaus™
30517981 |
1 kg |
8160.00 NOK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||||
16642322
|
Ohaus™
30517982 |
2 kg |
11995.00 NOK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
|||||
16652322
|
Ohaus™
30517983 |
5 kg |
20090.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
16662322
|
Ohaus™
30517984 |
10 kg |
28725.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
16672322
|
Ohaus™
30517985 |
20 kg |
50550.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
Description
- OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
- Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
- Weights ranging in masses from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
Specifications
No Certificate | |
Troemner Alloy 8 | |
Protective Case |