Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML E1
Adjusted to Meet or Exceed OIML E1 Tolerances
Brand:  Ohaus™ 30517984
28018.58 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16662322
  • 28725.00 NOK / Each
In Stock
Explore more special offers
Add to basket
Description
Description
  • OIML Class E1 offers high accuracy with tight tolerances and guaranteed uncertainty 1/3 of the tolerance
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish
  • Weights ranging in masses from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free, protecting your investment
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No Certificate
10 kg
Protective Case
Troemner Alloy 8