Learn More
EMP Entreprise Metallo Plastique™ Retention Tank with Platform
Includes grating and platform. EMP Entreprise Metallo Plastique™ Retention Tray with Platform prevents workfloors from becoming hazardous and slippery.
POA - 149.54€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11917695
![]() |
EMP Entreprise Metallo Plastique™
PFCP100006F |
100 L |
149.54€
Each |
Estimated Shipment: 11-09-2025
Log in to see stock. |
|||||
11927695
![]() |
EMP Entreprise Metallo Plastique™
PFCP200006F |
200 L |
POA
Each |
N/A | |||||
Description
- Constructed of 100% Polyethylene
- High chemical resistance
- High retention capacity
- Easy cleaning
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.