Learn More
EMP Entreprise Metallo Plastique™ Retention Tank with Platform
Includes grating and platform. EMP Entreprise Metallo Plastique™ Retention Tray with Platform prevents workfloors from becoming hazardous and slippery.
Brand:  EMP Entreprise Metallo Plastique™ PFCP100006F
Encompass
 View more versions of this product
Product Code. 11917695
  • 149.54€ / Each
Only null left
Estimated Shipment: 11-09-2025
Description
  • Constructed of 100% Polyethylene
  • High chemical resistance
  • High retention capacity
  • Easy cleaning
Specifications
Specifications
100 L
Certificates
Promotions
Promotions
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.