Learn More
OHAUS™ PVC Flask Clamp
OHAUS™ PVC Flask Clamp is designed to securely grip and position various sizes of flasks.
503.00 NOK - 1155.00 NOK
Specifications
Quantity | 1 |
---|---|
Type | Flask Clamp |
Material | PVC |
Color | Purple |
Jaw Type | Four Interlocking |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15712519
|
OHAUS™
30400099 |
125 mL Flask |
503.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
15722519
|
OHAUS™
30400100 |
250 mL Flask |
525.00 NOK
Each |
Estimated Shipment: 17-10-2025
Log in to see stock. |
|||||
15732519
|
OHAUS™
30400101 |
500 mL Flask |
605.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
15742519
|
OHAUS™
30400102 |
1000 mL Flask |
668.00 NOK
Each |
Estimated Shipment: 17-10-2025
Log in to see stock. |
|||||
15752519
|
OHAUS™
30400103 |
2000 mL Flask |
1155.00 NOK
Each |
In Stock
Log in to see stock. |
|||||
Description
- Constructed of PVC
- For use with flasks
- Designed to securely hold flask in place
Specifications
1 | |
PVC | |
Four Interlocking |
Flask Clamp | |
Purple |