Learn More
OHAUS™ PVC Flask Clamp
OHAUS™ PVC Flask Clamp is designed to securely grip and position various sizes of flasks.
Brand:  OHAUS™ 30400102
648.21 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15742519
  • 668.00 NOK / Each
Only null left
Estimated Shipment: 22-10-2025
Description
  • Constructed of PVC
  • For use with flasks
  • Designed to securely hold flask in place
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1
PVC
Flask Clamp PVC 1000mL
Four Interlocking
Flask Clamp
1000 mL Flask
Purple
5.66