missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ N20 PE Closures
Brand: Macherey-Nagel™ 70240
411.80 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Specifications
Transparent | |
Butyl beige / PTFE gray unassembled | |
100 Pk. | |
Closed Top Cap |
Butyl Rubber, Polyethylene, PTFE | |
9.1 mm | |
1.3 mm | |
Flat DIN Crimp Neck |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction