Learn More
Buerkle™ Laboratory Bottle PET Sterile
Sterile laboratory bottle for use in laboratories and chemical handling areas. Good product presentation due to crystal-clear PET. Space-saving and absolutely leakproof. Ideal for storing and packing liquids.
41.55 NOK - 42.92 NOK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
15722785
|
Buerkle™
0370-0125 |
125 mL |
41.55 NOK
Each |
Estimated Shipment: 12-09-2025
Log in to see stock. |
|||||
15477321
|
Buerkle™
0370-0250 |
250 mL |
41.55 NOK
Each |
Estimated Shipment: 08-09-2025
Log in to see stock. |
|||||
16423919
|
Buerkle™
0370-0500 |
500 mL |
42.92 NOK
Each |
Estimated Shipment: 15-09-2025
Log in to see stock. |
|||||
Description
- Sterile SAL 10-3 (according to ISO 11137)
- PET crystal-clear
- With tamper-proof closure (HDPE)
- With moulded-in raised graduation
Specifications
PET |