Learn More
Buerkle™ Laboratory Bottle PET Sterile
Sterile laboratory bottle for use in laboratories and chemical handling areas. Good product presentation due to crystal-clear PET. Space-saving and absolutely leakproof. Ideal for storing and packing liquids.
Brand:  Buerkle™ 0370-0500
38.58 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 16423919
  • 42.92 NOK / Each
Focus on Agri-Science Testing
Use promo code 24136 at checkout
View Available Promotions
Only null left
Estimated Shipment: 15-09-2025
Description
  • Sterile SAL 10-3 (according to ISO 11137)
  • PET crystal-clear
  • With tamper-proof closure (HDPE)
  • With moulded-in raised graduation
Specifications
Specifications
500 mL
PET