Learn More
O&M Halyard Quick Check Sterilization Wrap (H200)
Using Halyard Quick Check wraps is a fast, easy way to ensure instrument sterility is uncompromised.
1264.89 NOK - 3247.67 NOK
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | Meets NFPA Class 1 |
Color | Blue |
For Use With (Equipment) | Linen towels, packs and gowns, light basin sets and single instruments (3 to 6 lb.) |
Material | SMS Fabric (H200) |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
30274672
|
O&M
34167 |
30 x 30 cm (12 x 12 in.) |
1287.67 NOK
Pack of 480 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
30274775
|
O&M
34193 |
45 x 45 cm (18 x 18 in.) |
2016.97 NOK
Pack of 480 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
30274681
|
O&M
34174 |
60 x 60 cm (24 x 24 in.) |
1726.40 NOK
Pack of 240 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
30274744
|
O&M
|
76 x 76 cm (30 x 30 in.) |
1435.80 NOK
Pack of 144 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
30274674
|
O&M
34185 |
91 x 91 cm (36 x 36 in.) |
2222.09 NOK
Pack of 144 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
30274702
|
O&M
34186 |
101 x 101 cm (40 x 40 in.) |
2056.85 NOK
Pack of 120 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
30274730
|
O&M
34149 |
121 x 121 cm (48 x 48 in.) |
1264.89 NOK
Pack of 48 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
30274687
|
O&M
34187 |
137 x 137 cm (54 x 54 in.) |
1937.21 NOK
Pack of 48 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
30274781
|
O&M
|
137 x 182 cm (54 x 72 in.) |
3247.67 NOK
Pack of 48 |
Estimated Shipment: 16-06-2025
Log in to see stock. |
|||||
Description
- Inspect trays easier and faster than ever
- Superior microbial barrier protection with patented Powerguard technology
- Cleared for all of the same sterilization systems as Halyard Kimguard One-step Wrap
- Made from recyclable No. 5 polypropylene fabric
- Made in the USA
Specifications
Autoclavable | |
Blue | |
SMS Fabric (H200) | |
Yes |
Meets NFPA Class 1 | |
Linen towels, packs and gowns, light basin sets and single instruments (3 to 6 lb.) | |
Non-sterile | |
Sterilization Wrap |