missing translation for 'onlineSavingsMsg'
Learn More

GNMT Mouse anti-Human, Unconjugated, Clone: 1B5E3, Proteintech

Mouse Monoclonal Antibody

1696.32 NOK - 4796.50 NOK

Specifications

Antigen GNMT
Clone 1B5E3
Concentration 1000 μg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16809845
View Documents
Proteintech
67294-1-IG-20UL
20 μL
1696.32 NOK
20µL
Please to purchase this item. Need a web account? Register with us today!
16899835
View Documents
Proteintech
67294-1-IG-150UL
150 μL
4796.50 NOK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The GNMT antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNMT. This antibody recognizes human, mouse, pig, rat antigen. The GNMT antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

GNMT
1000 μg/mL
Monoclonal
Mouse
27232
Primary
1B5E3
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Human, Mouse, Rat, Pig
IgG1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.