Learn More
GNMT Mouse anti-Human, Unconjugated, Clone: 1B5E3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67294-1-IG-20UL
 View more versions of this product
Product Code. 16809845
  • 1696.32 NOK / 20µL
Description
The GNMT antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNMT. This antibody recognizes human, mouse, pig, rat antigen. The GNMT antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
GNMT
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, ELISA
1B5E3
Unconjugated
20 μL
27232
IgG1