Learn More
OHAUS™ Frontier™ 5000 Multi-Pro Series Bucket
Offering flexibility to maximize sample preparation.
Brand: OHAUS™ 83041013
4866.69 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Frontier™ Multi-Pro Series centrifuge accessory.
- Bucket
- 200 mL
- 2 pack
Specifications
83041011 | |
Bucket | |
Aluminum |
1 x 200 mL | |
With Cap | |
200 mL |