Learn More
OHAUS™ Frontier™ 5000 Multi-Pro Series Bucket
Offering flexibility to maximize sample preparation.
Brand:  OHAUS™ 83041012
4293.45 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 30130248
  • 4405.00 NOK / Pack of 2
Description
Frontier™ Multi-Pro Series centrifuge accessory.
  • Bucket
  • 200 mL
  • 2 pack
TRUSTED_SUSTAINABILITY
Specifications
Specifications
83041011
Bucket
Aluminum
1 x 200 mL
Without Cap
200 mL