Learn More
Lenz Laborglasintrumente Erlenmeyer Flasks With DIN Thread
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
2325.00 NOK - 2980.00 NOK
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Price | Quantity & Availability | |||||
10290050
|
Lenz Laborglasintrumente
03025149 |
85 mm |
2735.00 NOK
Pack of 10 |
Estimated Shipment: 08-08-2025
Log in to see stock. |
|||||
10755442
|
Lenz Laborglasintrumente
03025137 |
64 mm |
2325.00 NOK
Pack of 10 |
Estimated Shipment: 08-08-2025
Log in to see stock. |
|||||
10833581
|
Lenz Laborglasintrumente
03025158 |
105 mm |
2980.00 NOK
Pack of 10 |
Estimated Shipment: 08-08-2025
Log in to see stock. |
|||||
Description
- With DIN thread (GL) and screw cap (red)
- With scale (approx. volume /ml)
- With inscription label
- Material: DURAN™ (flask), PBT (cap)
Specifications
10/Pk. |