missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRCP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14348-1-AP-20UL
This item is not returnable.
View return policy
Specifications
CRCP | |
Polyclonal | |
Unconjugated | |
CRCP | |
CGRP RCP, CGRP receptor component, CGRPRCP, CRCP, HsC17, RCP, RCP9, RNA polymerase III subunit C9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114205, 12909, 27297 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35427, O75575, Q8VHM6 | |
CRCP | |
CRCP Fusion Protein Ag5689 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction