Learn More
Lenz Laborglasintrumente Cones
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
366.00 NOK - 1430.00 NOK
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Joint Size | Price | Quantity & Availability | |||||
10578271
|
Lenz Laborglasintrumente
01100260 |
NS 60/46 |
453.00 NOK
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10121521
|
Lenz Laborglasintrumente
01100271 |
NS 71/51 |
754.00 NOK
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10189690
|
Lenz Laborglasintrumente
01100207 |
NS 7/16 |
667.00 NOK
Pack of 10 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10254001
|
Lenz Laborglasintrumente
01100240 |
NS 40/38 |
1430.00 NOK
Pack of 10 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10450703
|
Lenz Laborglasintrumente
01100250 |
NS 50/42 |
366.00 NOK
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10493861
|
Lenz Laborglasintrumente
01100205 |
NS 5/13 |
924.00 NOK
Pack of 10 |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10528851
|
Lenz Laborglasintrumente
01100285 |
NS 85/55 |
1280.00 NOK
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
10569951
|
Lenz Laborglasintrumente
01100255 |
NS 55/44 |
396.00 NOK
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
|||||
Description
- Acc. to DIN 12249 form B and in addition to standard
- Conical ground cones
- Made of DURAN™ tubing
Specifications
Borosilicate Glass (3.3) |