Learn More
Lenz Laborglasintrumente Cones
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01100240
 View more versions of this product
Product Code. 10254001
  • 1420.00 SEK / Pack of 10
Only null left
Estimated Shipment: 24-09-2025
Description
  • Acc. to DIN 12249 form B and in addition to standard
  • Conical ground cones
  • Made of DURAN™ tubing
Specifications
Specifications
10/Pk.
2 mm
Borosilicate Glass (3.3)
NS 40/38
36 mm