Learn More
Endecotts™ Sieve Calibration Standards
Use for calibration of the woven wire mesh sieves. All reference samples supplied by Endecotts are provided with a Certificate of Calibration and are fully traceable to National Standards
2853.07 NOK - 3009.99 NOK
Product Code | Brand | Particle Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Price | Quantity & Availability | |||||
11924305
|
Endecotts™
ZSICSA-.212 |
212 μm |
3009.99 NOK
Pack of 5 |
Estimated Shipment: 27-11-2025
Log in to see stock. |
|||||
11964305
|
Endecotts™
ZSICSA-.500 |
500 μm |
2853.07 NOK
Pack of 5 |
Estimated Shipment: 27-11-2025
Log in to see stock. |
|||||
11954295
|
Endecotts™
ZSICSA-.053 |
53 μm |
3009.99 NOK
Pack of 5 |
Estimated Shipment: 27-11-2025
Log in to see stock. |
|||||
11934305
|
Endecotts™
ZSICSA-.250 |
250 μm |
2853.07 NOK
Pack of 5 |
Estimated Shipment: 27-11-2025
Log in to see stock. |
|||||
11944305
|
Endecotts™
ZSICSA-.300 |
300 μm |
2853.07 NOK
Pack of 5 |
Estimated Shipment: 27-11-2025
Log in to see stock. |
|||||
Description
- Calibrates sieves to an accuracy of ±1μm
- Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture.
- Traceable to the United Kingdom National Physical Laboratory
- Standards include a calibration chart.
Specifications
Soda-lime glass | |
Sieve |