Learn More
Endecotts™ Sieve Calibration Standards
Use for calibration of the woven wire mesh sieves. All reference samples supplied by Endecotts are provided with a Certificate of Calibration and are fully traceable to National Standards
Brand: Endecotts™ ZSICSA-.500
This item is not returnable.
View return policy
Description
- Calibrates sieves to an accuracy of ±1μm
- Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture.
- Traceable to the United Kingdom National Physical Laboratory
- Standards include a calibration chart.
Specifications
Calibration Ball Mesh | |
Sieve | |
500 μm |
Soda-lime glass | |
5/Pk. |