Learn More
Buffer Solution pH 4.01, Crison Instruments™
24.50€ - 31.40€
Specifications
pH | 4.01 |
---|---|
Solution Type | Buffer |
Formulation | Potassium Hydrogen Phthalate, Germicide |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12041462
|
Crison Instruments
LZW9460.99 |
125 mL |
24.50€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12802173
|
Crison Instruments
CR94660 |
1000 mL | N/A | N/A | N/A | ||||
Specifications
4.01 | |
Buffer | |
Potassium Hydrogen Phthalate, Germicide |