Learn More
Buffer Solution pH 4.01, Crison Instruments™
Click to view available options
Quantity:
125 mL
1000 mL
Specifications
Specifications
pH | 4.01 |
Quantity | 125 mL |
Solution Type | Buffer |
Formulation | Potassium Hydrogen Phthalate, Germicide |