missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol purple, MP Biomedicals™
549.00 NOK - 5270.00 NOK
Chemical Identifiers
CAS | 115-40-2 |
---|---|
Molecular Formula | C21H16Br2O5S |
Molecular Weight (g/mol) | 540.22 |
MDL Number | MFCD00011681 |
InChI Key | ABIUHPWEYMSGSR-UHFFFAOYSA-N |
Synonym | 5', 5”-dibromo-o-cresolsulfonephthalein |
PubChem CID | 8273 |
ChEBI | CHEBI:86154 |
IUPAC Name | 3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione |
SMILES | CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11440942
|
MP Biomedicals
0219508005 |
5 g |
549.00 NOK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450942
|
MP Biomedicals
0219508025 |
25 g |
1660.00 NOK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11460942
|
MP Biomedicals
0219508080 |
100 g |
5270.00 NOK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
115-40-2 | |
540.22 | |
ABIUHPWEYMSGSR-UHFFFAOYSA-N | |
8273 | |
3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione |
C21H16Br2O5S | |
MFCD00011681 | |
5', 5”-dibromo-o-cresolsulfonephthalein | |
CHEBI:86154 | |
CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1 |
Specifications
115-40-2 | |
MFCD00011681 | |
ABIUHPWEYMSGSR-UHFFFAOYSA-N | |
3,3-bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione | |
8273 | |
540.24 | |
Crystalline Powder |
C21H16Br2O5S | |
5', 5”-dibromo-o-cresolsulfonephthalein | |
CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1 | |
540.22 | |
CHEBI:86154 | |
Brown to Purple | |
Bromocresol purple |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title