missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromocresol purple, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Specifications
Specifications
Formula Weight | 540.24 |
Color | Brown to Purple |
Quantity | 25 g |
Physical Form | Crystalline Powder |
Chemical Name or Material | Bromocresol purple |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction