Learn More
Bismuth(III) isopropoxide, Thermo Scientific Chemicals
1396.00 NOK - 9145.00 NOK
Chemical Identifiers
CAS 15049-67-9
Molecular Formula C9H21BiO3
Molecular Weight (g/mol) 386.244
MDL Number MFCD00799065
InChI Key KNPRLIQQQKEOJN-UHFFFAOYSA-N
Synonym bismuth iii isopropoxide, triisopropoxybismuthine, acmc-20all6, bismuth 3+ tripropan-2-olate, bismuth 3+ tris propan-2-olate
PubChem CID 22648700
IUPAC Name bismuth;propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Bi+3]
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11304988
SDS View Documents
Thermo Scientific Alfa Aesar
043938.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11314988
SDS missing translation for 'viewDocuments'
Thermo Scientific Alfa Aesar
043938.06
5 g
9145.00 NOK
5g
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under Argon
Specifications
Chemical Identifiers
15049-67-9
386.244
KNPRLIQQQKEOJN-UHFFFAOYSA-N
22648700
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Bi+3]
C9H21BiO3
MFCD00799065
bismuth iii isopropoxide, triisopropoxybismuthine, acmc-20all6, bismuth 3+ tripropan-2-olate, bismuth 3+ tris propan-2-olate
bismuth;propan-2-olate
Specifications
15049-67-9
C9H21BiO3
MFCD00799065
Moisture Sensitive
KNPRLIQQQKEOJN-UHFFFAOYSA-N
bismuth;propan-2-olate
22648700
Powder
Gray
Bi[OCH(CH3)2]3
UN3181
bismuth iii isopropoxide, triisopropoxybismuthine, acmc-20all6, bismuth 3+ tripropan-2-olate, bismuth 3+ tris propan-2-olate
CC(C)[O-].CC(C)[O-].CC(C)[O-].[Bi+3]
386.244
386.24
Bismuth (III) isopropoxide
Videos
Safety and Handling
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P280g-P305+P351+P338-P337+P313
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL SALTS OF ORGANIC COMPOUNDS, FLAMMABLE, N.O.S.
missing translation for 'storageNote' :
  • Packaged under Argon
Product Identifier
  • Bismuth(III) isopropoxide
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.