missing translation for 'onlineSavingsMsg'
Learn More

Bismuth(III) isopropoxide, Thermo Scientific Chemicals

Product Code. 11304988
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15049-67-9
Molecular Formula C9H21BiO3
Molecular Weight (g/mol) 386.244
MDL Number MFCD00799065
InChI Key KNPRLIQQQKEOJN-UHFFFAOYSA-N
Synonym bismuth iii isopropoxide, triisopropoxybismuthine, acmc-20all6, bismuth 3+ tripropan-2-olate, bismuth 3+ tris propan-2-olate
PubChem CID 22648700
IUPAC Name bismuth;propan-2-olate
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].[Bi+3]

Specifications

Color Gray
Linear Formula Bi[OCH(CH3)2]3
Quantity 1 g
UN Number UN3181
Sensitivity Moisture Sensitive
Formula Weight 386.24
Physical Form Powder
Chemical Name or Material Bismuth (III) isopropoxide
compliance-icons compliance-icons
Product Identifier
  • Bismuth(III) isopropoxide
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H228-Flammable solid.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.