Learn More
Lenz Laborglasintrumente 3-Neck Adapters
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
701.00 NOK - 819.00 NOK
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | |||||
10376261
|
Lenz Laborglasintrumente
05015011 |
Cone 14/23 NS, Sockets 14/23 NS |
701.00 NOK
Each |
Estimated Shipment: 02-07-2025
Log in to see stock. |
|||||
10438982
|
Lenz Laborglasintrumente
05015023 |
Cone 24/29 NS, Sockets 19/26 NS |
760.00 NOK
Each |
Estimated Shipment: 27-06-2025
Log in to see stock. |
|||||
10448982
|
Lenz Laborglasintrumente
05015033 |
Cone 24/29 NS, Sockets 24/29 NS |
819.00 NOK
Each |
Estimated Shipment: 23-06-2025
Log in to see stock. |
|||||
Description
- With 3 NS sockets and 1 NS cone
- With two parallel necks and one angled neck (45°)
- Made of DURAN™ tubing
Specifications
Borosilicate Glass (3.3) |