Learn More
Lenz Laborglasintrumente 3-Neck Adapters
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05015011
 View more versions of this product
Product Code. 10376261
  • 701.00 NOK / Each
Estimated Shipment: 02-07-2025
Explore more special offers
Description
  • With 3 NS sockets and 1 NS cone
  • With two parallel necks and one angled neck (45°)
  • Made of DURAN™ tubing
Specifications
Specifications
Borosilicate Glass (3.3)
Cone 14/23 NS, Sockets 14/23 NS