Learn More
Zirconium Nickel powder, -325 mesh, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Chemical Name or Material | Zirconium Nickel powder |
Color | Gray |
Avg. Mol. Wt. or Mol. Wt. Range | Zr:Ni; 70:30 wt% |
MDL Number | MFCD00192582 |
UN Number | UN3089 |
Quantity | 25 g |
Mesh Size | −325 Mesh |
Sensitivity | Air Sensitive |
Form | Powder |