Learn More
ZBED1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26140-1-AP-150UL
 View more versions of this product
Product Code. 16894665
  • 4796.50 NOK / 150µL
Explore more special offers
Description
Description
ZBED1, also known as DREF, is the human homolog of the Drosophila protein DREF, a transcriptional regulatory factor required for expression of genes involved in DNA replication and cell proliferation. RNA interference (RNAi) experiments targeting ZBED1 RNA resulted in the inhibition of S phase entry and reduction of histone H1 mRNA in HeLa cells, suggesting that ZBED1 also plays a role in cell proliferation in human cells. At least 22 ribosomal proteins (RPs) possess ZBED1 binding sites in their promoters; anti-ZBED1 RNAi also resulted in decreased RP gene expression. Immunoprecipitation analyses have shown that ZBED1 self-associates in vivo via a carboxyl-terminal hATC domain, and this self-association is required for nuclear localization and DNA binding.
Specifications
Specifications
ZBED1
Polyclonal
Unconjugated
ZBED1
ALTE, DREF, dREF homolog, hDREF, KIAA0785, TRAMP, ZBED1
Rabbit
Antigen Affinity Chromatography
RUO
9189
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O96006
ZBED1
ZBED1 Fusion Protein Ag23578
150 μL
Primary
Human, Mouse