Learn More
Tenak™ Yellow Cardboard Cryobox
Cryobox made of water-repellent cardboard for long-lasting durability.
Brand: Tenak™ TE22088
2050.56 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Constructed of fibreboard/cardboard
- Water-repellent for longer durability
- Can be used with a large range of grid dividers
Specifications
Yellow | |
Square | |
133 x 133 x 130 mm |
Cardboard | |
Cryo Box |