Learn More
WNT2 Mouse anti-Human, Mouse, Rat, Clone: 3H7B10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66656-1-IG-20UL
This item is not returnable.
View return policy
Description
WNT2 is a ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.Specifications
WNT2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P09544, P21552 | |
WNT2 | |
WNT2 Fusion Protein Ag25185 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunofluorescence, Western Blot | |
3H7B10 | |
Unconjugated | |
WNT2 | |
Int 1 like protein 1, Int 1 related protein, INT1L1, IRP, Protein Wnt 2, WNT2 | |
Mouse | |
Protein G | |
RUO | |
114487, 22413, 7472 | |
-20°C | |