Learn More
BRAND™ Wide Neck PFA Bottle
Shop All Brand Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
2000 mL
Description
- PFA screw cap with a molded sealing ring for a positive seal
- Autoclavable
- Easy to clean
- Withstands temperatures from -200 to 260°C
Specifications
Specifications
Capacity (Metric) | 500 mL |
Description | Wide Neck |
Material | PFA |
Thread Size (Metric) | S40 |
Closure Type | Screw Cap |