Learn More
DWK Life Sciences Wheaton™ Black Phenolic Screw Caps Without Liner
Offers superior torque retention and excellent sealing characteristics
Brand: DWK Life Sciences 239853
1120.43 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Poly–Seal liner is manufactured from low density polyethylene
- Screw Cap
- Phenolic
- Black
It is recommended that liner be tested prior to use for leak resistant applications
Specifications
Black | |
500 Cs. | |
22-350 | |
651905/907 Vacules and stopper |
Phenolic Resin | |
39 mm | |
ISO 9001 |