Learn More
DWK Life Sciences Wheaton™ E-Z Seals With PTFE/Grey Butyl Liner
Brand: DWK Life Sciences 224235-01
3079.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
PTFE/Butyl | |
Aluminum | |
1000 Cs. | |
11 mm |
Natural | |
E-Z Seal | |
Open Top Cap |